Assay Details
Gene Symbol: Angptl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199115.2 | NP_954546.1 | 3 - 4 | 686 | 83 | |
GenBank mRNA | AY393999.1 | - | 3 - 4 | 546 | 83 | |
BC078944.1 | - | 3 - 4 | 686 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
