Assay Details
Gene Symbol: Stx16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108610.1 | NP_001102080.1 | 4 - 5 | 486 | 102 | |
XM_006235693.2 | XP_006235755.1 | 6 - 7 | 1414 | 102 | ||
XM_006235694.2 | XP_006235756.1 | 5 - 6 | 1402 | 102 | ||
XM_006235695.2 | XP_006235757.1 | 6 - 7 | 1363 | 102 | ||
XM_006235696.2 | XP_006235758.1 | 5 - 6 | 1350 | 102 | ||
GenBank mRNA | BC168980.1 | - | 3 - 4 | 340 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
