Assay Details
Gene Symbol: Cdk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191861.1 | NP_001178790.1 | 1 - 2 | 232 | 111 | |
XM_006236019.2 | XP_006236081.1 | 2 - 3 | 696 | 111 | ||
XM_006236020.2 | XP_006236082.1 | 2 - 3 | 630 | 111 | ||
XM_006236021.1 | XP_006236083.1 | 2 - 3 | 572 | 111 | ||
XM_008762679.1 | XP_008760901.1 | 2 - 3 | 547 | 111 | ||
GenBank mRNA | AF352168.1 | - | 1 - 2 | 160 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
