Assay Details
Gene Symbol: Cdk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191861.1 | NP_001178790.1 | 2 - 3 | 364 | 142 | |
XM_006236019.2 | XP_006236081.1 | 3 - 4 | 828 | 142 | ||
XM_006236020.2 | XP_006236082.1 | 3 - 4 | 762 | 142 | ||
XM_006236021.1 | XP_006236083.1 | 3 - 4 | 704 | 142 | ||
XM_008762679.1 | XP_008760901.1 | 3 - 4 | 679 | 142 | ||
GenBank mRNA | AF352168.1 | - | 2 - 3 | 292 | 142 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
