Assay Details
Gene Symbol: Cdk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191861.1 | NP_001178790.1 | 3 - 4 | 540 | 87 | |
XM_006236019.2 | XP_006236081.1 | 4 - 5 | 1004 | 87 | ||
XM_006236020.2 | XP_006236082.1 | 4 - 5 | 938 | 87 | ||
XM_006236021.1 | XP_006236083.1 | 4 - 5 | 880 | 87 | ||
XM_008762679.1 | XP_008760901.1 | 4 - 5 | 855 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
