Assay Details
Gene Symbol: Sri | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128190.1 | NP_001121662.1 | 5 - 6 | 412 | 77 | |
XM_006236007.2 | XP_006236069.1 | 5 - 6 | 445 | 77 | ||
GenBank mRNA | BC158845.1 | - | 5 - 6 | 412 | 77 | |
FQ220011.1 | - | 5 - 6 | 424 | 77 | ||
FQ226061.1 | - | 5 - 6 | 422 | 77 | ||
FQ226813.1 | - | 5 - 6 | 422 | 77 | ||
FQ227927.1 | - | 5 - 6 | 407 | 77 | ||
FQ229846.1 | - | 5 - 6 | 77 | |||
FQ232170.1 | - | 5 - 6 | 422 | 77 | ||
FQ234730.1 | - | 5 - 6 | 424 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
