Assay Details
Gene Symbol: Ccdc132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173511.1 | NP_001166982.1 | 25 - 26 | 2541 | 101 | |
XM_006236037.2 | XP_006236099.1 | 25 - 26 | 2572 | 101 | ||
XM_008762704.1 | XP_008760926.1 | 25 - 26 | 2575 | 101 | ||
GenBank mRNA | BC091288.1 | - | 10 - 11 | 1136 | 101 |
Gene Symbol: LOC100911994 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749697.3 | XP_003749745.2 | 5 - 6 | 1799 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
