Assay Details
Gene Symbol: Bet1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019251.2 | NP_062124.1 | 2 - 3 | 226 | 180 | |
GenBank mRNA | BC062408.1 | - | 2 - 3 | 226 | 180 | |
FQ223263.1 | - | 2 - 3 | 301 | 180 | ||
U42755.1 | - | 2 - 3 | 270 | 180 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
