Assay Details
Gene Symbol: Cd63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017125.3 | NP_058821.1 | 4 - 5 | 389 | 120 | |
XM_008765024.1 | XP_008763246.1 | 4 - 5 | 451 | 120 | ||
XM_008765025.1 | XP_008763247.1 | 4 - 5 | 654 | 120 | ||
GenBank mRNA | BC063173.1 | - | 4 - 5 | 392 | 6921594 | 120 |
FQ214468.1 | - | 4 - 5 | 456 | 120 | ||
FQ221333.1 | - | 4 - 5 | 393 | 120 | ||
FQ222038.1 | - | 4 - 5 | 372 | 120 | ||
FQ230116.1 | - | 4 - 5 | 360 | 120 | ||
X61654.1 | - | 4 - 5 | 389 | 120 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
