Assay Details
Gene Symbol: Nabp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034939.1 | NP_001030111.2 | 3 - 4 | 694 | 114 | |
NM_001244819.1 | NP_001231748.1 | 4 - 5 | 417 | 114 | ||
XM_006240769.2 | XP_006240831.1 | 4 - 5 | 658 | 114 | ||
XM_006240770.2 | XP_006240832.1 | 4 - 5 | 513 | 114 | ||
XM_006240771.2 | XP_006240833.1 | 4 - 5 | 508 | 114 | ||
XM_008765029.1 | XP_008763251.1 | 4 - 5 | 673 | 114 | ||
GenBank mRNA | BC104710.1 | - | 3 - 4 | 694 | 114 | |
DV214819.1 | - | 4 - 5 | 417 | 114 | ||
FQ219805.1 | - | 4 - 5 | 405 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
