Assay Details
Gene Symbol: Cdk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199501.1 | NP_955795.1 | 2 - 3 | 342 | 117 | |
XM_006240778.1 | XP_006240840.1 | 2 - 3 | 418 | 117 | ||
GenBank mRNA | BC061832.1 | - | 2 - 3 | 342 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
