Assay Details
Gene Symbol: Cdk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199501.1 | NP_955795.1 | 5 - 6 | 734 | 67 | |
GenBank mRNA | BC061832.1 | - | 5 - 6 | 734 | 67 | |
D28753.1 | - | 6 - 7 | 770 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
