Assay Details
Gene Symbol: G6pd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017006.2 | NP_058702.1 | 9 - 10 | 1157 | 66 | |
GenBank mRNA | BC081820.1 | - | 9 - 10 | 1070 | 7108556 | 66 |
X07467.1 | - | 9 - 10 | 1083 | 66 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
