Assay Details
Gene Symbol: Dkc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133419.1 | NP_596910.1 | 1 - 2 | 107 | 87 | |
XM_008773596.1 | XP_008771818.1 | 1 - 2 | 508 | 87 | ||
GenBank mRNA | BC087600.1 | - | 1 - 2 | 66 | 87 | |
BC099832.1 | - | 1 - 2 | 65 | 87 | ||
Z34922.1 | - | 1 - 2 | 107 | 87 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC099832.1, NM_133419.1, XM_008773596.1, Z34922.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
