Assay Details
Gene Symbol: Fam3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109324.1 | NP_001102794.1 | 4 - 5 | 548 | 62 | |
XM_006229603.2 | XP_006229665.1 | 3 - 4 | 705 | 62 | ||
GenBank mRNA | BC169081.1 | - | 3 - 4 | 612 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
