Assay Details
Gene Symbol: Fam3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109324.1 | NP_001102794.1 | 5 - 6 | 666 | 67 | |
XM_006229603.2 | XP_006229665.1 | 4 - 5 | 823 | 67 | ||
XM_006229604.2 | XP_006229666.1 | 4 - 5 | 3235 | 67 | ||
XM_006229605.2 | XP_006229667.1 | 5 - 6 | 2548 | 67 | ||
GenBank mRNA | BC169081.1 | - | 4 - 5 | 730 | 5621009 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
