Assay Details
Gene Symbol: Bcap31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004224.1 | NP_001004224.1 | 2 - 3 | 182 | 69 | |
XM_006229556.2 | XP_006229618.1 | 1 - 2 | 139 | 69 | ||
XM_006229557.2 | XP_006229619.1 | 2 - 3 | 256 | 69 | ||
GenBank mRNA | BC079182.1 | - | 2 - 3 | 182 | 7130880 | 69 |
FQ219566.1 | - | 2 - 3 | 173 | 69 | ||
FQ219790.1 | - | 2 - 3 | 181 | 69 | ||
FQ230132.1 | - | 2 - 3 | 173 | 69 | ||
FQ230136.1 | - | 2 - 3 | 173 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
