Assay Details
Gene Symbol: Bcap31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004224.1 | NP_001004224.1 | 6 - 7 | 681 | 89 | |
XM_006229556.2 | XP_006229618.1 | 5 - 6 | 638 | 89 | ||
XM_006229557.2 | XP_006229619.1 | 6 - 7 | 755 | 89 | ||
GenBank mRNA | BC079182.1 | - | 6 - 7 | 681 | 7130880 | 89 |
FQ219566.1 | - | 6 - 7 | 672 | 89 | ||
FQ230132.1 | - | 6 - 7 | 672 | 89 | ||
FQ230136.1 | - | 6 - 7 | 672 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
