Assay Details
Gene Symbol: LOC100910130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749779.3 | XP_003749827.1 | 12 - 13 | 1118 | 65 |
Gene Symbol: Fam50a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170573.1 | NP_001164044.1 | 12 - 13 | 1134 | 65 | |
GenBank mRNA | BC168887.1 | - | 10 - 11 | 727 | 65 | |
FQ229851.1 | - | 12 - 13 | 1069 | 65 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
