Assay Details
Gene Symbol: Mrs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024001.1 | NP_076491.1 | 10 - 11 | 1272 | 97 | |
GenBank mRNA | AF288289.1 | - | 10 - 11 | 1272 | 97 | |
AF288290.1 | - | 6 - 7 | 723 | 97 | ||
AF288291.1 | - | 6 - 7 | 725 | 97 | ||
BC083554.1 | - | 10 - 11 | 1265 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
