Assay Details
Gene Symbol: Mrs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024001.1 | NP_076491.1 | 2 - 3 | 312 | 136 | |
XM_006253937.2 | XP_006253999.1 | 2 - 3 | 430 | 136 | ||
GenBank mRNA | AF288289.1 | - | 2 - 3 | 312 | 136 | |
BC083554.1 | - | 2 - 3 | 288 | 136 | ||
BC098916.1 | - | 2 - 3 | 287 | 136 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
