Assay Details
Gene Symbol: Scgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201561.1 | NP_963855.1 | 5 - 6 | 392 | 120 | |
XM_008771653.1 | XP_008769875.1 | 6 - 7 | 798 | 120 | ||
GenBank mRNA | AY513659.1 | - | 5 - 6 | 392 | 120 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
