Assay Details
Gene Symbol: Cmah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024273.1 | NP_001019444.1 | 14 - 15 | 1825 | 98 | |
GenBank mRNA | BC095843.1 | - | 14 - 15 | 1825 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P127026 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454865 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849816 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 772290 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777613 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170026 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710190 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781108 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822050 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966597 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387700 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 51548 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936764 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707432 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972122 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729832 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268380 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40761 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721659 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549641 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396411 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574104 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546545 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778249 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909418 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481848 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914915 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622514 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619212 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492506 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275871 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197190 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897156 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427675 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101178 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715590 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91966 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59029 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167988 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27804 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85957 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76424 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156965 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P127026 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P127026 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P127026 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P552681 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 3816 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 75110 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 71734 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 68141 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10798425 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10798430 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10798423 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10798427 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10798429 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10798422 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10798424 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10798421 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10798426 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10798428 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10798420 | - | - |
Rat Genome 230 2.0 Array | 1380432_PM_at | - | - |
Rat Genome 230 2.0 Array | 1380432_at | - | - |
Rat Genome 230 2.0 Array | 1384476_at | - | - |
Rat Genome 230 2.0 Array | 1384476_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1380432_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1384476_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1384476_at | - | - |
Affymetrix HT Rat Focus Array | 1380432_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1700012168.rn.1 | TC1700001774.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1700012169.rn.1 | TC1700001774.rn.1 | RTA TC |
Rat Expression Set 230 | 1380432_at | - | - |
Rat Expression Set 230 | 1384476_PM_at | - | - |
Rat Expression Set 230 | 1384476_at | - | - |
Rat Expression Set 230 | 1380432_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10798420 | - | - |
Rat Genome U34 Set | rc_AI014030_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21015996 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1354723 | - | - |
