Assay Details
Gene Symbol: Hist1h2ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022643.1 | NP_072169.1 | 1 | 47 | 91 | |
GenBank mRNA | M18045.1 | - | 1 | 47 | 91 | |
M18046.1 | - | 1 | 47 | 91 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the M18045.1, M18046.1, NM_022643.1 transcripts.
This assay is in the 5'UTR region of the M18045.1, M18046.1, NM_022643.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
