Assay Details
Gene Symbol: Prl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012629.1 | NP_036761.1 | 2 - 3 | 388 | 121 | |
XM_006253912.2 | XP_006253974.1 | 2 - 3 | 280 | 121 | ||
GenBank mRNA | AF022935.1 | - | 2 - 3 | 388 | 121 | |
BC166892.1 | - | 2 - 3 | 253 | 121 | ||
BC168729.1 | - | 2 - 3 | 259 | 121 | ||
S46794.1 | - | 1 - 2 | 124 | 121 | ||
V01249.1 | - | 2 - 3 | 253 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
