Assay Details
Gene Symbol: Prl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012629.1 | NP_036761.1 | 4 - 5 | 667 | 91 | |
XM_006253912.2 | XP_006253974.1 | 4 - 5 | 559 | 91 | ||
GenBank mRNA | AF022935.1 | - | 4 - 5 | 667 | 91 | |
BC166892.1 | - | 4 - 5 | 532 | 91 | ||
BC168729.1 | - | 4 - 5 | 538 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
