Assay Details
Gene Symbol: Marf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133421.2 | NP_596912.1 | 12 - 13 | 2817 | 175 | |
XM_003750779.3 | XP_003750827.1 | 12 - 13 | 3864 | 175 | ||
XM_006245854.2 | XP_006245916.1 | 12 - 13 | 2749 | 175 | ||
XM_006245855.2 | XP_006245917.1 | 12 - 13 | 2643 | 175 | ||
XM_006245856.2 | XP_006245918.1 | 12 - 13 | 2254 | 175 | ||
XM_008767442.1 | XP_008765664.1 | 11 - 12 | 3298 | 175 | ||
GenBank mRNA | AB012133.1 | - | 12 - 13 | 2817 | 175 |
Gene Symbol: LOC103689943 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006245686.2 | XP_006245748.1 | 12 - 13 | 3865 | 175 | |
XM_006245687.2 | XP_006245749.1 | 12 - 13 | 2749 | 175 | ||
XM_006245688.1 | XP_006245750.1 | 12 - 13 | 2643 | 175 | ||
XM_006245689.2 | XP_006245751.1 | 12 - 13 | 2254 | 175 | ||
XM_008767447.1 | XP_008765669.1 | 12 - 13 | 2791 | 175 | ||
XM_008767448.1 | XP_008765670.1 | 11 - 12 | 3298 | 175 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
