Assay Details
Gene Symbol: Fam63b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008766411.1 | XP_008764633.1 | 4 - 5 | 1326 | 99 | |
XM_343420.8 | XP_343421.3 | 4 - 5 | 1318 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
