Assay Details
Gene Symbol: Aqp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022960.2 | NP_075249.1 | 6 - 7 | 703 | 143 | |
XM_006243362.1 | XP_006243424.1 | 5 - 6 | 808 | 143 | ||
XM_008766336.1 | XP_008764558.1 | 2 - 3 | 509 | 143 | ||
GenBank mRNA | AF016406.1 | - | 2 - 3 | 482 | 143 | |
BC085731.1 | - | 6 - 7 | 703 | 143 | ||
FQ209455.1 | - | 2 - 3 | 504 | 143 | ||
FQ211101.1 | - | 2 - 3 | 424 | 143 | ||
FQ212527.1 | - | 2 - 3 | 387 | 143 | ||
FQ214432.1 | - | 5 - 6 | 813 | 143 | ||
FQ218988.1 | - | 2 - 3 | 507 | 143 | ||
FQ219175.1 | - | 2 - 3 | 424 | 143 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
