Assay Details
Gene Symbol: Ccnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009470.1 | NP_001009470.1 | 2 - 3 | 217 | 121 | |
GenBank mRNA | BC088212.1 | - | 2 - 3 | 217 | 121 | |
BC097952.1 | - | 2 - 3 | 241 | 121 | ||
FQ228772.1 | - | 2 - 3 | 231 | 121 | ||
FQ234119.1 | - | 2 - 3 | 231 | 121 |
Gene Symbol: LOC100364016 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008758651.1 | XP_008756873.1 | 1 | 251 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
