Assay Details
Gene Symbol: LOC100364016 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008758651.1 | XP_008756873.1 | 1 | 1074 | 118 |
Gene Symbol: Ccnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009470.1 | NP_001009470.1 | 7 - 8 | 1040 | 118 | |
GenBank mRNA | BC088212.1 | - | 7 - 8 | 1040 | 118 | |
BC097952.1 | - | 7 - 8 | 1064 | 118 | ||
FQ228772.1 | - | 7 - 8 | 1054 | 118 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
