Assay Details
Gene Symbol: Lipc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012597.2 | NP_036729.2 | 3 - 4 | 551 | 81 | |
XM_006243359.2 | XP_006243421.1 | 5 - 6 | 654 | 81 | ||
XM_006243360.2 | XP_006243422.1 | 4 - 5 | 606 | 81 | ||
GenBank mRNA | BC088160.1 | - | 3 - 4 | 551 | 81 | |
M16235.1 | - | 3 - 4 | 473 | 81 | ||
X17366.1 | - | 3 - 4 | 461 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
