Assay Details
Gene Symbol: Lipc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012597.2 | NP_036729.2 | 4 - 5 | 669 | 139 | |
XM_006243359.2 | XP_006243421.1 | 6 - 7 | 772 | 139 | ||
XM_006243360.2 | XP_006243422.1 | 5 - 6 | 724 | 139 | ||
XM_006243361.2 | XP_006243423.1 | 4 - 5 | 541 | 139 | ||
GenBank mRNA | BC088160.1 | - | 4 - 5 | 669 | 139 | |
M16235.1 | - | 4 - 5 | 591 | 139 | ||
X17366.1 | - | 4 - 5 | 579 | 139 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
