Assay Details
Gene Symbol: Lipc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012597.2 | NP_036729.2 | 5 - 6 | 903 | 146 | |
XM_006243359.2 | XP_006243421.1 | 7 - 8 | 1006 | 146 | ||
XM_006243360.2 | XP_006243422.1 | 6 - 7 | 958 | 146 | ||
XM_006243361.2 | XP_006243423.1 | 5 - 6 | 775 | 146 | ||
GenBank mRNA | BC088160.1 | - | 5 - 6 | 903 | 146 | |
M16235.1 | - | 5 - 6 | 825 | 146 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
