Assay Details
Gene Symbol: Lipc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012597.2 | NP_036729.2 | 8 - 9 | 1479 | 128 | |
XM_006243359.2 | XP_006243421.1 | 10 - 11 | 1582 | 128 | ||
XM_006243360.2 | XP_006243422.1 | 9 - 10 | 1534 | 128 | ||
XM_006243361.2 | XP_006243423.1 | 8 - 9 | 1351 | 128 | ||
GenBank mRNA | BC088160.1 | - | 8 - 9 | 1479 | 128 | |
M16235.1 | - | 8 - 9 | 1401 | 128 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
liver development
fatty acid metabolic process
fatty acid biosynthetic process
triglyceride metabolic process
phosphatidylserine metabolic process
circadian rhythm
cholesterol metabolic process
response to xenobiotic stimulus
response to hormone
response to carbohydrate
response to acetate
response to inorganic substance
response to organic cyclic compound
heparan sulfate proteoglycan biosynthetic process
triglyceride catabolic process
cholesterol transport
response to nutrient levels
response to magnesium ion
cellular response to hormone stimulus
response to lipid
chylomicron remodeling
very-low-density lipoprotein particle remodeling
low-density lipoprotein particle remodeling
high-density lipoprotein particle remodeling
chylomicron remnant clearance
low-density lipoprotein particle clearance
cholesterol homeostasis
response to amino acid
response to peptide hormone
phosphatidylethanolamine metabolic process
neutral lipid catabolic process
phosphatidylcholine metabolic process
phosphatidic acid metabolic process
glycerophospholipid catabolic process
response to copper ion
developmental growth
protein complex oligomerization
response to glucocorticoid
response to calcium ion
triglyceride homeostasis
response to fatty acid
regulation of plasma lipoprotein particle levels
lysophospholipase activity
triglyceride lipase activity
heparin binding
lipid binding
phospholipase A1 activity
lipase activity
acyltransferase activity
low-density lipoprotein particle binding
apolipoprotein binding
chylomicron binding
heparan sulfate proteoglycan binding
acylglycerol lipase activity
carboxylic ester hydrolase activity
response to hypoxia
liver development
fatty acid metabolic process
fatty acid biosynthetic process
triglyceride metabolic process
phosphatidylserine metabolic process
circadian rhythm
cholesterol metabolic process
response to xenobiotic stimulus
response to hormone
response to carbohydrate
response to acetate
response to inorganic substance
response to organic cyclic compound
heparan sulfate proteoglycan biosynthetic process
triglyceride catabolic process
cholesterol transport
response to nutrient levels
response to magnesium ion
cellular response to hormone stimulus
response to lipid
chylomicron remodeling
very-low-density lipoprotein particle remodeling
low-density lipoprotein particle remodeling
high-density lipoprotein particle remodeling
chylomicron remnant clearance
low-density lipoprotein particle clearance
cholesterol homeostasis
response to amino acid
response to peptide hormone
phosphatidylethanolamine metabolic process
neutral lipid catabolic process
phosphatidylcholine metabolic process
phosphatidic acid metabolic process
glycerophospholipid catabolic process
response to copper ion
developmental growth
protein complex oligomerization
response to glucocorticoid
response to calcium ion
triglyceride homeostasis
response to fatty acid
regulation of plasma lipoprotein particle levels
lysophospholipase activity
triglyceride lipase activity
heparin binding
lipid binding
phospholipase A1 activity
lipase activity
acyltransferase activity
low-density lipoprotein particle binding
apolipoprotein binding
chylomicron binding
heparan sulfate proteoglycan binding
acylglycerol lipase activity
carboxylic ester hydrolase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P313838 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964695 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379754 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404471 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28624 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719650 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41102 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332565 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201859 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829663 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253552 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771743 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299212 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891730 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890292 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308645 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665684 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492898 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297867 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529544 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556132 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375158 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905405 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732523 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573937 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624760 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244872 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801085 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86747 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716804 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336810 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 301588 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787514 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618967 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741086 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971743 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955220 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773617 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871306 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617726 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521357 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963342 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953666 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614293 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721957 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648213 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371468 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470019 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797525 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235015 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949847 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294350 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743946 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912340 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649641 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949983 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71178 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 554298 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920840 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915461 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799993 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446920 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764983 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762050 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614472 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281123 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914954 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781952 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586164 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524524 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920681 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6012 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84368 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921649 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683894 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444419 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182317 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784733 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601117 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310634 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61944 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 5610 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684175 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753581 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88317 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613408 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373955 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796259 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420499 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42612 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63143 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686835 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416183 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12873 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81040 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27063 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58266 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75537 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31418 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131674 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99015 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27421 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94762 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104557 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146163 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P313838 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P313838 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P313838 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P313838 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 60573 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 89544 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 1508 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 73889 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 61790 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 49829 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 35029 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 78741 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 33444 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10918566 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918565 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918562 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918568 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918563 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918564 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918561 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918567 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918560 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10918559 | - | - |
Rat Genome 230 2.0 Array | 1369701_at | - | - |
Rat Genome 230 2.0 Array | 1369701_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1369701_at | - | - |
Affymetrix HT Rat Focus Array | 1369701_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0800023222.rn.1 | TC0800002874.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0800023221.rn.1 | TC0800002874.rn.1 | RTA TC |
Rat Expression Set 230 | 1369701_at | - | - |
Rat Expression Set 230 | 1369701_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918559 | - | - |
Rat Genome U34 Set | M16235_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21300824 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1367184 | - | - |
RatRef-12 Expression BeadChip | ILMN_50862 | - | - |
