Assay Details
Gene Symbol: E2f5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_574892.6 | XP_574892.1 | 1 | 116 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the XM_574892.6 transcripts.
This assay is in the 5'UTR region of the XM_574892.6 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
