Assay Details
Gene Symbol: Cdc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108352.1 | NP_001101822.1 | 10 - 11 | 1351 | 69 | |
XM_006250570.2 | XP_006250632.1 | 8 - 9 | 1087 | 69 | ||
XM_008769940.1 | XP_008768162.1 | 8 - 9 | 1064 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
