Assay Details
Gene Symbol: Cdc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108352.1 | NP_001101822.1 | 11 - 12 | 1502 | 65 | |
XM_006250570.2 | XP_006250632.1 | 9 - 10 | 1238 | 65 | ||
XM_008769940.1 | XP_008768162.1 | 9 - 10 | 1215 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
