Assay Details
Gene Symbol: Rpl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031099.1 | NP_112361.1 | 1 - 2 | 129 | 101 | |
XM_006250585.2 | XP_006250647.1 | 1 - 2 | 110 | 101 | ||
GenBank mRNA | BC060561.1 | - | 1 - 2 | 76 | 101 | |
FQ212710.1 | - | 1 - 2 | 86 | 101 | ||
FQ214281.1 | - | 1 - 2 | 77 | 101 | ||
FQ214493.1 | - | 1 - 2 | 77 | 101 | ||
FQ215029.1 | - | 1 - 2 | 78 | 101 | ||
FQ219244.1 | - | 1 - 2 | 75 | 101 | ||
FQ219787.1 | - | 1 - 2 | 77 | 101 | ||
FQ220161.1 | - | 1 - 2 | 77 | 101 | ||
FQ220170.1 | - | 1 - 2 | 77 | 101 | ||
FQ220205.1 | - | 1 - 2 | 77 | 101 | ||
FQ220219.1 | - | 1 - 2 | 77 | 101 | ||
FQ227439.1 | - | 1 - 2 | 77 | 101 | ||
FQ228473.1 | - | 1 - 2 | 77 | 101 | ||
FQ229451.1 | - | 1 - 2 | 78 | 101 | ||
FQ229816.1 | - | 1 - 2 | 75 | 101 | ||
FQ229964.1 | - | 1 - 2 | 79 | 101 | ||
FQ229979.1 | - | 1 - 2 | 77 | 101 | ||
FQ230017.1 | - | 1 - 2 | 75 | 101 | ||
FQ230061.1 | - | 1 - 2 | 80 | 101 | ||
FQ230102.1 | - | 1 - 2 | 77 | 101 | ||
FQ230128.1 | - | 1 - 2 | 105 | 101 | ||
FQ230161.1 | - | 1 - 2 | 103 | 101 | ||
FQ230794.1 | - | 1 - 2 | 80 | 101 | ||
FQ231729.1 | - | 1 - 2 | 75 | 101 | ||
FQ232540.1 | - | 1 - 2 | 77 | 101 | ||
FQ233314.1 | - | 1 - 2 | 102 | 101 | ||
FQ234171.1 | - | 1 - 2 | 107 | 101 | ||
X06148.1 | - | 1 - 2 | 129 | 101 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC060561.1, NM_031099.1, X06148.1, XM_006250585.2 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
