Assay Details
Gene Symbol: Tmlhe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133387.2 | NP_596878.2 | 5 - 6 | 750 | 71 | |
XM_006255855.1 | XP_006255917.1 | 4 - 5 | 759 | 71 | ||
GenBank mRNA | AF374406.1 | - | 3 - 4 | 591 | 71 | |
BC085923.1 | - | 5 - 6 | 750 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
