Assay Details
Gene Symbol: Nlrc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772331.1 | XP_008770553.1 | 8 - 9 | 3761 | 70 |
Gene Symbol: LOC102553150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006255134.2 | XP_006255196.1 | 1 - 2 | 686 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006255134.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
