Assay Details
Gene Symbol: Got2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013177.2 | NP_037309.1 | 2 - 3 | 359 | 131 | |
GenBank mRNA | BC061792.1 | - | 2 - 3 | 307 | 5597941 | 131 |
FM056810.1 | - | 2 - 3 | 359 | 131 | ||
M18467.1 | - | 2 - 3 | 330 | 131 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
