Assay Details
Gene Symbol: Ciapin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007689.1 | NP_001007690.1 | 6 - 7 | 931 | 65 | |
XM_006255106.2 | XP_006255168.1 | 6 - 7 | 1018 | 65 | ||
XM_006255107.2 | XP_006255169.1 | 6 - 7 | 730 | 65 | ||
XM_006255108.2 | XP_006255170.1 | 6 - 7 | 733 | 65 | ||
GenBank mRNA | BC083753.1 | - | 6 - 7 | 931 | 65 | |
FQ211546.1 | - | 6 - 7 | 715 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
