Assay Details
Gene Symbol: Hmgxb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107412.1 | NP_001100882.1 | 3 - 4 | 320 | 62 | |
XM_006255141.2 | XP_006255203.1 | 2 - 3 | 4738 | 62 | ||
XM_006255143.1 | XP_006255205.1 | 3 - 4 | 322 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
