Assay Details
Gene Symbol: Hmgxb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107412.1 | NP_001100882.1 | 5 - 6 | 1335 | 114 | |
XM_006255141.2 | XP_006255203.1 | 4 - 5 | 5753 | 114 | ||
XM_006255143.1 | XP_006255205.1 | 5 - 6 | 1337 | 114 | ||
XM_006255144.1 | XP_006255206.1 | 3 - 4 | 1142 | 114 | ||
XM_008772312.1 | XP_008770534.1 | 3 - 4 | 1320 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
