Assay Details
Gene Symbol: Colec12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025721.1 | NP_001020892.1 | 9 - 10 | 2291 | 125 | |
GenBank mRNA | BC097495.1 | - | 9 - 10 | 2291 | 7461783 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P020466 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418271 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 476733 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149278 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701806 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921079 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832970 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 611132 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947040 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128221 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734992 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785663 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107104 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7024 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347133 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790502 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812688 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427830 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240270 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776032 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109697 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57886 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684557 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534610 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763906 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193907 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332177 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122772 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596485 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796228 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203580 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36472 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13682 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721364 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634284 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717263 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210565 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390379 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823988 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72484 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556729 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244777 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915647 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661034 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784600 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774508 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225067 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610939 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163017 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164490 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952815 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536323 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44374 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731203 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800592 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886996 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11296 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563442 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814339 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616528 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636484 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671788 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920439 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665115 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354007 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557870 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297323 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302907 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486459 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66935 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258626 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455482 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699119 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325994 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18898 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263852 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312356 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542843 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673747 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882008 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737376 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74236 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681547 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365111 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952151 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736769 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653845 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609328 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711546 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417352 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969409 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226038 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518245 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789223 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183905 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36060 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395534 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620886 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187158 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342621 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751279 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854569 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640740 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805133 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269717 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714017 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457083 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692507 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358155 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815431 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310286 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27885 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604501 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641531 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801134 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380044 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479380 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176705 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68729 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27984 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23813 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105596 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145696 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121860 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155636 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138666 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107164 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75254 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108850 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76722 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10377 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70016 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6245 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P020466 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P020466 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P020466 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P598365 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P288029 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 49234 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 8575 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 43191 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 25982 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 63259 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 80079 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 70574 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 68066 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 16071 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 82889 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 74625 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 58498 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 69180 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 86669 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 62690 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 44032 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10800094 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800095 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800093 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800089 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800097 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800092 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800096 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800090 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800091 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10800088 | - | - |
Rat Genome 230 2.0 Array | 1372818_at | - | - |
Rat Genome 230 2.0 Array | 1372818_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1372818_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1372818_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1800000076.rn.1 | TC1800000010.rn.1 | RTA TC |
Rat Expression Set 230 | 1372818_PM_at | - | - |
Rat Expression Set 230 | 1372818_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800088 | - | - |
Rat Genome U34 Set | rc_AI180192_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21337559 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1359457 | - | - |
