Assay Details
Gene Symbol: Abhd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106162.1 | NP_001099632.1 | 8 - 9 | 1106 | 124 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P23203 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393750 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776430 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634796 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963485 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289302 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923056 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120502 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764757 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742056 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322836 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344841 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426973 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636305 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591997 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127025 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969178 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408032 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21673 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59312 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389926 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752747 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433566 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829793 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497224 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107164 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27539 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690733 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314111 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124427 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59358 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88574 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164949 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P23203 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P23203 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P23203 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P23203 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 30388 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 27871 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 57616 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10803210 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10803215 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10803212 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10803209 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10803213 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10803211 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10803208 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10803214 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10803207 | - | - |
Rat Genome 230 2.0 Array | 1381773_PM_at | - | - |
Rat Genome 230 2.0 Array | 1381773_at | - | - |
Rat Genome 230 2.0 Array | 1382137_PM_at | - | - |
Rat Genome 230 2.0 Array | 1382137_at | - | - |
Affymetrix HT Rat Focus Array | 1382137_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1382137_at | - | - |
Affymetrix HT Rat Focus Array | 1381773_at | - | - |
Affymetrix HT Rat Focus Array | 1381773_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1800007255.rn.1 | TC1800001017.rn.1 | RTA TC |
Rat Expression Set 230 | 1382137_at | - | - |
Rat Expression Set 230 | 1382137_PM_at | - | - |
Rat Expression Set 230 | 1381773_PM_at | - | - |
Rat Expression Set 230 | 1381773_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10803207 | - | - |
Rat Genome U34 Set | rc_AI227905_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21651188 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1364145 | - | - |
