Assay Details
Gene Symbol: Mrps18b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212534.2 | NP_997699.2 | 3 - 4 | 306 | 83 | |
GenBank mRNA | BC166875.1 | - | 3 - 4 | 288 | 83 | |
FM036709.1 | - | 3 - 4 | 306 | 83 | ||
FQ215273.1 | - | 3 - 4 | 279 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
