Assay Details
Gene Symbol: Gtf2h4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212501.2 | NP_997666.1 | 2 - 3 | 311 | 94 | |
XM_006255920.2 | XP_006255982.1 | 2 - 3 | 360 | 94 | ||
GenBank mRNA | BC083703.1 | - | 2 - 3 | 269 | 94 | |
BC127469.1 | - | 2 - 3 | 283 | 94 | ||
CK472761.1 | - | 2 - 3 | 311 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
