Assay Details
Gene Symbol: Atat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212498.1 | NP_997663.1 | 3 - 4 | 271 | 139 | |
XM_006255948.2 | XP_006256010.1 | 3 - 4 | 306 | 139 | ||
XM_006255950.2 | XP_006256012.1 | 3 - 4 | 306 | 139 | ||
XM_006255951.2 | XP_006256013.1 | 3 - 4 | 306 | 139 | ||
XM_008772731.1 | XP_008770953.1 | 3 - 4 | 306 | 139 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
